missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prox1 Antibody (5G10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-30045SS
This item is not returnable.
View return policy
Description
Prox1 Monoclonal specifically detects Prox1 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Whole-Mount, CyTOF-ready.
Specifications
| Prox1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry Whole-Mount reported in scientific literature (PMID 24654984), CyTOF-ready | |
| Q92786, Q92786, P48437 | |
| PROX1 | |
| The highly conserved Homeo and Prospero domain of human Prox1. [UniProt# Q92786] | |
| 0.025 mL | |
| Vision | |
| 5629 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 5G10 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction