missing translation for 'onlineSavingsMsg'
Learn More

Protriptyline hydrochloride, MedChemExpress

Product Code. 16404517
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 16404517

Brand: Medchem Express HYB0949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Protriptyline hydrochloride is a tricyclic antidepressant (TCA), specifically a secondary amine, for the treatment of depression and ADHD.

Protriptyline hydrochloride is a tricyclic antidepressant (TCA), specifically a secondary amine, for the treatment of depression and ADHD.

Specifications

Chemical Name or Material Protriptyline hydrochloride
Physical Form Solid
CAS 1225-55-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C19H22ClN
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 250 mg/mL (833.78 mM; Need ultrasonic) ∣H2O : 100 mg/mL (333.51 mM; Need ultrasonic)
SMILES CNCCCC1C2=CC=CC=C2C=CC3=CC=CC=C13.[H]Cl
Molecular Weight (g/mol) 299.84
Formula Weight 299.84
Percent Purity 99.23%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.