missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protriptyline.HCl, 1mg/ml in Methanol (as free base), Lipomed
Description
Protriptyline.HCl, 1mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Protriptyline HCl |
| CAS | 1225-55-4 |
| Analyte or Component Names | Protriptyline HCl |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H21 N . Cl H |
| Concentration | 1 mg/mL |
| Synonym | N-Nethyl-5 H-dibenzo[a,d]cycloheptene-5-propanamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, Concordin, MK-240, Maximed, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride, N-Methyl-5 H-dibenzo[a,d]cycloheptene-5-propylamine hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CNCCCC1c2ccccc2C=Cc3ccccc13 |
| IUPAC Name | 3-(11 H-dibenzo[2,1-b:1',2'-e][7 ]annulen-11-yl)-N-methylpropan-1-amine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?