missing translation for 'onlineSavingsMsg'
Learn More

Protosappanin A, MedChemExpress

Product Code. 16437527
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16437527 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16437527 Supplier Medchem Express Supplier No. HY113573
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Protosappanin A (PTA), an immunosuppressive ingredient and major biphenyl compound isolated from Caesalpinia sappan L, suppresses JAK2/STAT3-dependent inflammation pathway through down-regulating the phosphorylation of JAK2 and STAT3.

Protosappanin A (PTA), an immunosuppressive ingredient and major biphenyl compound isolated from Caesalpinia sappan L, suppresses JAK2/STAT3-dependent inflammation pathway through down-regulating the phosphorylation of JAK2 and STAT3.

Specifications

Chemical Name or Material Protosappanin A
CAS 102036-28-2
Color White
Purity Grade Notes Research
Molecular Formula C15H12O5
Quantity 5 mg
Synonym PTA
Solubility Information DMSO : 250 mg/mL (918.27 mM; Need ultrasonic)
SMILES O=C1CC2=CC(O)=C(O)C=C2C3=CC=C(O)C=C3OC1
Molecular Weight (g/mol) 272.25
Formula Weight 272.25
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.