missing translation for 'onlineSavingsMsg'
Learn More

Protoporphyrin IX, MedChemExpress

Product Code. 16428706
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428706 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428706 Supplier Medchem Express Supplier No. HYB1247100mg
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Protoporphyrin IX is the final intermediate in the heme biosynthetic pathway.

Protoporphyrin IX is the final intermediate in the heme biosynthetic pathway.

Specifications

Chemical Name or Material Protoporphyrin IX
CAS 553-12-8
Color Dark Brown
Purity Grade Notes Research
Molecular Formula C34H34N4O4
Quantity 100 mg
Solubility Information DMSO : 2.08 mg/mL (3.70 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble)
SMILES O=C(O)CCC1=C2/C=C3C(CCC(O)=O)=C(C)C(/C=C(N/4)/C(C)=C(C=C)C4=C\C5=N/C(C(C=C)=C5C)=C\C(N2)=C1C)=N/3
Molecular Weight (g/mol) 562.66
Formula Weight 562.66
Percent Purity 95.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.