missing translation for 'onlineSavingsMsg'
Learn More

Protoporphyrin-9, TRC

Product Code. 30136248
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136248 100 mg 100mg
30136950 1 g 1g
30009214 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136248 Supplier Toronto Research Chemicals Supplier No. TRCP839138100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-12-8
Molecular Formula C34H34N4O4
Molecular Weight (g/mol) 562.66
Synonym Protoporphyrin IX, 7,12-Diethenyl-3,8,13,17-tetramethyl-21H,23H-porphine-2,18-dipropanoic Acid, Kammerer's Porphyrin, Levulan, NSC 177389, NSC 2632, Ooporphyrin, Protoporphyrin
IUPAC Name 3-[(1Z,4Z,9Z,15Z)-18-(2-carboxyethyl)-7,12-bis(ethenyl)-3,8,13,17-tetramethyl-21,23-dihydroporphyrin-2-yl]propanoic acid
SMILES C=CC1=C(C)C2=CC3=NC(=CC4=C(CCC(=O)O)C(=C(C=C5C(=C(C)C(=N5)C=C1N2)C=C)N4)C)C(=C3C)CCC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C34H34N4O4/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25/h7-8,13-16,35,38H,1-2,9-12H2,3-6H3,(H,39,40)(H,41,42)/b25-13-,26-13-,27-14-,28-15-,29-14-,30-15-,31-16-,32-16-
Formula Weight 562.258
Chemical Name or Material Protoporphyrin-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.