missing translation for 'onlineSavingsMsg'
Learn More

Protogenin Antibody [PE], Novus Biologicals™

Product Code. 30736806 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736806 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736806 Supplier Novus Biologicals Supplier No. AF6795P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Protogenin Polyclonal antibody specifically detects Protogenin in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protogenin
Applications Immunohistochemistry
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias FLJ12795, FLJ13221, FLJ25756, IGDCC5, immunoglobulin superfamily, DCC subclass, member 5, Protein Shen-Dan, protogenin, protogenin homolog (Gallus gallus), shen-dan
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Protegenin, Phe36-Ala952, Accession # Q2VWP7
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283659
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.