missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Protodioscin

Product Code. 16479108
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479108 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479108 Supplier MedChemExpress Supplier No. HYN0799
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Protodioscin, a major steroidal saponin in dioscoreae rhizome, has been shown to exhibit multiple biological actions, such as anti-hyperlipidemia, anti-cancer, sexual effects and cardiovascular properties.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 1049.2 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 55056-80-9
Product Type Natural Products
Purity 98.0%
Solubility H2O : 50 mg/mL (47.66 mM∣Need ultrasonic) ∣DMSO : 50 mg/mL (47.66 mM∣Need ultrasonic)
Target Endogenous Metabolite
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.