missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protocatechualdehyde, MedChemExpress
Description
Protocatechualdehyde (Catechaldehyde), a natural polyphenol compound isolated from the roots of radix Salviae Miltiorrhizae, is associated with a wide variety of biological activities and has been widely used in medicine as an antioxidant, anti-aging, an antibacterial and anti-inflammatory agent.
Specifications
Specifications
| Chemical Name or Material | Protocatechualdehyde |
| Physical Form | Solid |
| CAS | 139-85-5 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C7H6O3 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | Catechaldehyde Protocatechuic aldehyde Rancinamycin IV |
| Solubility Information | DMSO : ≥ 50 mg/mL (362.00 mM) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?