missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Protocadherin gamma A8 Monoclonal antibody specifically detects Protocadherin gamma A8 in Human samples. It is validated for Western Blot, ELISA, ELISA, KnockDown
Specifications
Specifications
| Antigen | Protocadherin gamma A8 |
| Applications | Western Blot, ELISA, Sandwich ELISA, KnockDown |
| Classification | Monoclonal |
| Clone | 1C11 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_114477 |
| Gene Alias | KIAA0327PCDH-GAMMA-A8, PCDH-gamma-A8, protocadherin gamma subfamily A, 8, protocadherin gamma-A8 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?