missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protocadherin-19 Antibody (921614) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8626L
This item is not returnable.
View return policy
Description
Protocadherin-19 Monoclonal antibody specifically detects Protocadherin-19 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Protocadherin-19 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 921614 | |
| Flow Cytometry, CyTOF-ready | |
| EFMR;EIEE9;KIAA1313;PCDH19 | |
| HEK293 human embryonic kidney cell line transfected with human Protocadherin-19, Met1-Ser678, Accession # Q8TAB3 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction