missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protocadherin-10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19719
This item is not returnable.
View return policy
Description
Protocadherin-10 Polyclonal specifically detects Protocadherin-10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Protocadherin-10 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| DKFZp761O2023, KIAA1400PCDH19, MGC133344, OL-PCDH, ortholog of OL-pcdh, protocadherin 10, protocadherin-10 | |
| Rabbit | |
| 113 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q9P2E7 | |
| PCDH10 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human PCDH10. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 57575 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction