missing translation for 'onlineSavingsMsg'
Learn More

Novex™ ProtoArray™ Human Protein Microarray

Product Code. 15429764
Change view
Click to view available options
Number of Arrays:
1 array
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Number of Arrays unitSize
15429764 1 array Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15429764 Supplier Novex™ Supplier No. PAH0525101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ProtoArray™ Human Protein Microarray

The ProtoArray™ Human Protein Microarray is an advanced, high-content, functional protein microarray that enables rapid profiling of thousands of biochemical interactions in as little as one day. Perform a variety of assays, including biomarker identification, drug target discovery, enzyme substrate identification, antibody specificity profiling, and protein-protein interaction studies. Please email us for pricing information on bulk array purchases and custom services.

A Discovery Study Bundle is also available (Cat. No. PAH05251020). This bundle includes 20 ProtoArray™ Human Protein Microarrays.

Benefits of the ProtoArray™ Human Protein Microarray:

  • Target over over 9,000 unique human proteins selected from multiple gene families
  • Screen against native proteins purified and printed under non-denaturing conditions
  • Profile interactions in key biological pathways, such as cell signaling, inflammation, and apoptosis
  • Target key proteins, including cytokines, chemokines, and growth factors
  • Interrogate hundreds of known cancer-associated proteins for autoimmune responses

    Study Thousands of Human Proteins At Once
    This latest version (5.1) of the ProtoArray™ Human Protein Microarray contains over 9,000 unique human proteins selected from multiple gene families. This array can be used for a broad range of applications including:

  • Identifying novel protein biomarkers specific for disease states, including transplantation, cancers, and autoimmune diseases
  • Discovering and identifying drug target pathways
  • Profiling antibody specificity for research and therapeutic antibody development
  • Mapping protein-protein interactions important to biochemical pathways

    Utilizes Active Proteins from the Invitrogen™ Ultimate™ ORF Collection
    The ProtoArray™ Human Protein Microarray is designed for maximum protein functionality. The proteins are produced from clones selected from the Invitrogen™ Ultimate™ ORF Collection, the first and only clone resource to offer full-insert sequenced clones in a Gateway™ vector. Clones are expressed as N-terminal GST fusion proteins using a baculovirus expression system. All proteins are purified under nondenaturing conditions and printed at 4°C to preserve native protein structure.

    Have Confidence in Your Results
    To help ensure confidence in your results, proteins are arrayed in duplicate on a 1 inch by 3 inch glass slide along with many controls for background, labeling, and detection. The glass slide is an ultra-thin nitrocellulose-coated slide specifically selected for its low background binding. Each slide is labeled with a unique bar code for easy tracking. ProtoArray™ protein microarrays are easily read with most commercially available fluorescent microarray scanners.

    ProtoArray™ Prospector v5.2 is a free data analysis software tool that provides rapid interpretation of supported application experiments using ProtoArray™ microarrays. Learn more about ProtoArray™ Prospector v5.2.

    Related Links
    Learn more about ProtoArray™ protein microarrays.
    Learn more about Life Technologies' Ultimate™ ORF Collection.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Type ProtoArray™ Human Protein Microarray
    Number of Arrays 1 array
    Content And Storage -20°C
    Form Solid
    For Use With (Application) Protein arrays, protein assays and analysis, protein biology, protein microarrays, protein-protein interactions
    Includes One microarray with over 9,000 unique human proteins arrayed in duplicate on a 1 inch x 3 inch nitrocellulose-coated glass slide
    Quantity 1 Array

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.