missing translation for 'onlineSavingsMsg'
Learn More

PROTO-1, MedChemExpress

Product Code. 16477506
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477506 100 mg 100mg
16470048 5 mg 5mg
16413597 50 mg 50mg
16407578 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477506 Supplier Medchem Express Supplier No. HY18673

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PROTO-1 shows significant protection of hair cells, with HC50(concentration that would produce 50% hair cell survival) of 1 (mu)M-10 (mu)M (1 (mu)M≤HC50≤10 (mu)M).

PROTO-1 shows significant protection of hair cells, with HC50(concentration that would produce 50% hair cell survival) of 1 (mu)M-10 (mu)M (1 (mu)M≤HC50≤10 (mu)M).

Specifications

Chemical Name or Material PROTO-1
Physical Form Solid
CAS 312951-85-2
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H19ClN4O2S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 125 mg/mL (329.92 mM; Need ultrasonic)
SMILES O=C(C1=C(NC(NC2=CC=C(Cl)C=C2)=O)SC3=C1CCN(CC)C3)N
Molecular Weight (g/mol) 378.88
Formula Weight 378.88
Percent Purity 98.99%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.