missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protirelin, Mikromol™
Description
Protirelin
Specifications
Specifications
| Chemical Name or Material | D-His-Protirelin |
| CAS | 24305-27-9 |
| Analyte or Component Names | D-His-Protirelin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H22 N6 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Protirelin, 5-Oxo-l-prolyl-l-histidyl-l-prolinamide, L-Prolinamide, 5-oxo-L-prolyl-L-histidyl-, Thyrotropin-releasing factor (8CI), (Pyro)-L-glutamic acid-L-histidine-L-proline-NH2, 252: PN: WO2010033207 SEQID: 378 claimed protein, 83: PN: US20090175821 PAGE: 403 claimed protein, Abbott 38579, Antepan, FDA 1725, L-Pyroglutamyl-L-histidyl-L-prolinamide, L-Pyroglutamyl-L-histidyl-L-proline amide, Lopremone, Prem, Protirelin, Relefact TRH, Rifathyroin, Rifotironin, Ro 8-6270/9, Stimu TSH, Synthetic TRF, Synthetic TRH, Synthetic TSH-releasing factor, Synthetic TSH-releasing hormone, Synthetic thyrotropin-releasing factor, Synthetic thyrotropin-releasing hormone, TRF, TRH, TSH-RF, TSH-releasing factor, TSH-releasing hormone, Thypinone, Thyrefact, Thyroid releasing hormone, Thyroid-stimulating hormone-releasing factor, Thyroliberin, Thyrotropic hormone-releasing factor, Thyrotropic hormone-releasing hormone, Thyrotropic releasing hormone, Thyrotropic-releasing factor, Thyrotropin-releasing hormone |
| InChI Formula | InChI=1S/C16H22N6O4/c17-14(24)12-2-1-5-22(12)16(26)11(6-9-7-18-8-19-9)21-15(25)10-3-4-13(23)20-10/h7-8,10-12H,1-6H2,(H2,17,24)(H,18,19)(H,20,23)(H,21,25)/t10-,11-,12-/m0/s1 |
| SMILES | NC(=O)[C@@H]1CCCN1C(=O)[C@H](Cc2c[nH]cn2)NC(=O)[C@@H]3CCC(=O)N3 |
| IUPAC Name | (2S)-N-[(2S)-1-[(2S)-2-carbamoylpyrrolidin-1-yl]-3-(1H-imidazol-4-yl)-1-oxopropan-2-yl]-5-oxopyrrolidine-2-carboxamide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?