missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Protino Ni-TED 1000 kit for His-tag Protein Purification in Midi format

Product Code. 10031822
Change view
Click to view available options
:
No. of Reactions: 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10031822 No. of Reactions: 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10031822 Supplier Macherey-Nagel™ Supplier No. 745110.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The matrix of choice for highest protein purity in Midi format.

Protino™ NI-TED, the purer alternative for His-tag protein purification
  • Chelating group TED, one protein binding site
  • High stability against reducing/chelating agents
  • Low metal leaching
  • High protein purity
  • High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
  • Dry resin, storage at room temperature
Protino™ NI-TED packed columns
  • Ready to use pre-packed columns filled with dry Protino™ Ni-TED resin
  • Buffers included
Protino™ NI-TED resin
  • Dry bulk matrix
  • Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink well - highest flexibility
  • Batch procedure, gravity flow column chromatography, FPLC™ applications
  • Empty Protino™ columns available for use with Protino™ Ni-TED resin
  • Empty 96 well receiver plates available for use with Protino™ Ni-TED resin For purification of polyhistidine-tagged proteins; batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.

Specifications

Bed Volume 500 μL
Binding Capacity 2.5mg
Binding Property Batch
Kit Contents Buffers
Matrix Macroporous Silica
Quantity 1 Kit
For Use With (Application) For Purification of Polyhistidine-Tagged Proteins, Batch Binding, Gravity Flow Column Chromatography, MPLC/FPLC Applications
Type Ni-TED Protein Purification Kit
Ligand Type TED (tris-carboxymethyl ethylene diamine)
compliance-icons
Product Identifier
  • Protino Ni-TED 1000 packed columns (5)
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: imidazole, monosodium phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.