missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Protino™ Ni-NTA 5 mL FPLC™ Columns

Product Code. p-9281014
Click to view available options
Quantity:
5 mL
5 x 5 mL
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy

Product Code. 11982552

Brand: Macherey-Nagel™ 745415.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The method of choice for His-tag protein purification in a ready-to-use 5 mL FPCL columns.

  • Target: Protein
  • Technology: Immobilized metal ion affinity chromatography
  • Matrix: 6% beaded agarose (cross-linked), precharged with Ni2+
  • Bead size: 45 to 165 μm
  • Binding capacity: 250 mg
  • Enable fast and convenient purification of recombinant polyhistidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC)
  • Proteins from any expression system can be purified under native or denaturing

Specifications

For Use With Purification of His-tag proteins
Ligand Type NTA (nitrilotriacetic acid)
Quantity 5 mL
Temperature Store at 4 to 8°C
Type Protein solution
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.