missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Prothrombin Fragment 1/2 Monoclonal Antibody (D1), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA550944

 View more versions of this product

Product Code. 17938932

  • £161.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Prothrombin Fragment 1/2 is a protein fragment that is produced by the cleavage of prothrombin, a blood clotting protein. It is a small peptide consisting of the first half of the prothrombin molecule and is involved in the formation of blood clots. Prothrombin Fragment 1/2 is used as a biomarker for the diagnosis and monitoring of various blood clotting disorders.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Prothrombin Fragment 1/2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
P00734, P18292
F2
Recombinant Human F1+2 protein, Ala44-Arg327 (Accession #P00734).
20 μL
Primary
Human, Rat
Antibody
IgG2b κ
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
D1
Unconjugated
F2
prothrombin fragment 1.2; Prothrombin fragment 1+2; prothrombin fragment F1.2; Prothrombin fragment F1+2
Mouse
Protein A/G
RUO
2147, 29251
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.