missing translation for 'onlineSavingsMsg'
Learn More

Prothionamide, MedChemExpress

Product Code. 16468226
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16468226

Brand: Medchem Express HYB0306

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Protionamide (or prothionamide) is a drug used in the treatment of tuberculosis; has also been tested for use in the treatment of leprosy.

Protionamide (or prothionamide) is a drug used in the treatment of tuberculosis

Specifications

Chemical Name or Material Prothionamide
Physical Form Solid
CAS 14222-60-7
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H12N2S
For Use With (Application) Neuroscience-Neuromodulation
Synonym Protionamide
Solubility Information DMSO : 100 mg/mL (554.72 mM; Need ultrasonic)
SMILES S=C(C1=CC(CCC)=NC=C1)N
Molecular Weight (g/mol) 180.27
Formula Weight 180.27
Percent Purity 99.27%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.