Learn More
Tocris Bioscience™ Proteome Profiler Rat XL Cytokine Array
Product Code. 18084632
Click to view available options
:
Proteome Profiler Rat XL Cytokine Array
Product Code. 18084632
Brand: Tocris Bioscience™ ARY030
For parallel determination of relative levels of selected rat cytokines.
The Proteome Profiler Rat XL Cytokine Array contains 4 membranes-each spotted in duplicate with 79 different cytokine antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Description Four membranes-each spotted in duplicate with 79 different cytokine antibodies
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Kit Contents Array Buffer, Chemi Reagent, Detection Antibody Cocktail, Rat XL Cytokine Array, Rat XL Cytokine Array nitrocellulose membranes, Rectangular four-well multi-dish, Streptavidin-HRP, Transparency Overlay Template, Wash Buffer Concentrate (25X)
Product Type Capture Antigen ELISA kit
For Laboratory Research Use Only