Learn More
R&D Systems™ Proteome Profiler Human Protease Array Kit
GREENER_CHOICE
Product Code. 18027631
Product Code. 18027631
Brand: R&D Systems™ ARY021B
The Proteome Profiler Human Protease Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Human Protease Array Kit contains 4 membranes - each spotted in duplicate with 35 different protease antibodies.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit
Kit Contents 4 Array Membranes, 4-Well Multi-Dish, Array Buffers, Wash Buffer, Detection Antibody Cocktail, Streptavidin-HRP, Chemiluminescent Detection Reagents, Transparency Overlay Template, Detailed Protocol