missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Proteoglycans Joint Cartilage, Native Protein
Click to view available options
Quantity:
1mg
Unit Size:
Each
Description
Proteoglycans Joint Cartilage (human) Native Protein, RP-77524, from Invitrogen.
Human Joint Cartilage Extract - Proteoglycans are purified from human articulate cartilage using Guanidine- HCl extraction, pepsin digestion, ethanol precipitation, potassium-acetate extraction, and chromatography. RP-77524 is native Proteoglycans Joint Cartilage protein purified from human samples. RP-77524 may be used as a control with corresponding anti-Proteoglycans Joint Cartilage antibodies.
Specifications
Specifications
| For Use With (Application) | Control |
| Formulation | protein with no preservative |
| Name | Human Proteoglycans Joint Cartilage |
| Quantity | 1mg |
| Source | Human |
| Storage Requirements | 4°C |
| Regulatory Status | RUO |
| Common Name | Human Proteoglycans Joint Cartilage |
| Specific Reactivity | Human |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction