missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Proteinase K from Tritirachium album

Product Code. p-4605381 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
10 mg
25 mg
100 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
25mg
500mg
5mg
This item is not returnable. View return policy

Product Code. 11456391

Brand: MP Biomedicals™ 0219398110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly active stable endopeptidase with a broad spectrum of action

Proteinase K is used in digestion of proteins from brain tissue samples for prions in Transmissible Spongiform Encephalopathies (TSE) research. Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA, as well as in removal of endotoxins bound to cationic proteins such as lysozyme and ribonuclease A and in determination of enzyme localization on membranes.

  • Grade: Molecular biology reagent
  • Specific Activity: >30 mAnson u/mg
  • Presentation: White powder
  • Isoelectric Point: 8.9 (Lit.)
  • Extinction Coefficient: E1%= 14.2 (280 nm,10 mM NaCl and 5 mM CaCl2, pH 8.0) (Lit.)
  • pH: 7.5 -12.00 (denatured hemoglobin as substrate) (Lit.)
  • Soluble in water

One Anson unit is the amount of enzyme which will liberate 1 °mole of folin-positive amino acids per minnute at pH 7.5 and 35°C using hemoglobin as substrat

Isolation of DNA and RNA

TRUSTED_SUSTAINABILITY

Specifications

Color White
Grade Molecular Biology
pH 7.5 to 12
For Use With (Application) Useful for the proteolytic inactivation of Nucleases during the isolation of DNA and RNA
Content And Storage 4°C
Source Tritirachium album
Quantity 10 mg
Product Type Proteinase K
Molecular Weight (g/mol) 30000
Inhibitors Diisopropylfluorophosphate or Phenylmethane Sulfonyl Fluoride, AEBSF and Trypsin
Form Lyophilized Powder
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Proteinase K, Molecular Biology Reagent
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Proteinase K
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.