missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Proteinase K, FS, for food safety testing

Product Code. 15206545 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1.25 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15206545 1.25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15206545 Supplier Applied Biosystems™ Supplier No. 4480715

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Proteinase K, FS, for food safety testing

Proteinase K, FS from the fungus Engyodontium album is a non-specific serine protease that is useful for general digestion of proteins. Proteinase K, FS is supplied specifically for use in food and environmental testing workflows. It is typically used in conjunction with Lysis Buffer, FS (available separately) to lyse microbial organisms in order to prepare the microbial DNA for downstream analysis by real-time PCR.

Proteinase K, FS can be used as a part of both the Pathatrix™ Auto Instrument and RapidFinder™ Direct Lysis workflows.

When used as a part of the Pathatrix™ Auto Instrument workflow, one bottle of Proteinase K, FS is sufficient for 1250 reactions.

When used as a part of the RapidFinder™ Direct Lysis workflow, one bottle of Proteinase K, FS is sufficient for 100 reactions.

Proteinase K, FS is supplied in 50% glycerol at a concentration of 20 mg/ml.

Unit Definition: One unit liberates 1μmol of Folin-positive amino acids, measured as tyrosine, at 37°C, pH 7.5, using denatured bovine hemoglobin as the substrate.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Food safety testing
Content And Storage -20°C
Quantity 1.25 mL
Product Type Proteinase K

For Testing of Food and Environmental Samples Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.