missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Lyophilized, Proteinase K

Product Code. 15880918
Change view
Click to view available options
Quantity:
100 mg (2 x 50 mg)
30 mg
Storage Buffer:
1.8mL Proteinase Buffer PB
8mL Proteinase Buffer PB
Unit Size:
100mg
30mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Storage Buffer unitSize
15880918 30 mg 1.8mL Proteinase Buffer PB 30mg
11912312 100 mg (2 x 50 mg) 8mL Proteinase Buffer PB 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15880918 Supplier Macherey-Nagel™ Supplier No. 740506.30

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Delivered with a stabilizing solution for enhanced stability. Macherey-Nagel™ Proteinase K is lyophilized proteinase K supplied in pre-measured vials for easy preparation of 20mg/mL stock solution. It can be used with NucleoSpin™ and NucleoBond™ kits.

Proteinase K is necessary for the isolation of DNA or simultaneous RNA / DNA isolation for DNA viruses (e.g. HBV) that are more difficult to lyse. Proteinase K treatment is also recommended for viral RNA isolation when viscous samples have to be processed (e.g., sputum samples).

  • Delivered with stabilizing solution to enable storage for up to six months at -20°C
  • No decrease of proteinase activity when dissolved in the buffer compared to dissolving in water
  • Lyophilized mataerial is supplied in precisely measured vials, to which buffer solution is added to prepare 20mg/mL stock solution
  • Suitable for use with the NucleoSpin™ RNA Virus F kit and other NucleoSpin™ and NucleoBond™ kits, and also suitable for use in most other applications

Specifications

Concentration 20 mg/mL
Color White
Components Buffer solution
For Use With (Application) Viral RNA and DNA isolation, including: HCV, HBV, HAV, HIV, HSV, HPV, VZV, EBV, parvovirus B19, H5N1
For Use With (Equipment) NucleoSpin™ and NucleoBond™ kits
Storage Buffer 1.8mL Proteinase Buffer PB
Packaging Vial
Content And Storage Proteinase and Buffer, separately: room temperature (18–25°C) for at least 1 year
Prepared Stock Solution: −20°C for up to 6 months
Source Tritirachium album
Quantity 30 mg
Product Type Proteinase K
Form Lyophilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Proteinase K
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.