missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Lyophilized, Proteinase K
Description
Proteinase K is necessary for the isolation of DNA or simultaneous RNA / DNA isolation for DNA viruses (e.g. HBV) that are more difficult to lyse. Proteinase K treatment is also recommended for viral RNA isolation when viscous samples have to be processed (e.g., sputum samples).
- Delivered with stabilizing solution to enable storage for up to six months at -20°C
- No decrease of proteinase activity when dissolved in the buffer compared to dissolving in water
- Lyophilized mataerial is supplied in precisely measured vials, to which buffer solution is added to prepare 20mg/mL stock solution
- Suitable for use with the NucleoSpin™ RNA Virus F kit and other NucleoSpin™ and NucleoBond™ kits, and also suitable for use in most other applications
Specifications
Specifications
| Concentration | 20 mg/mL |
| Color | White |
| Components | Buffer solution |
| For Use With (Application) | Viral RNA and DNA isolation, including: HCV, HBV, HAV, HIV, HSV, HPV, VZV, EBV, parvovirus B19, H5N1 |
| For Use With (Equipment) | NucleoSpin™ and NucleoBond™ kits |
| Storage Buffer | 8mL Proteinase Buffer PB |
| Packaging | Vial |
| Content And Storage | Proteinase and Buffer, separately: room temperature (18–25°C) for at least 1 year Prepared Stock Solution: −20°C for up to 6 months |
| Source | Tritirachium album |
| Quantity | 100 mg (2 x 50 mg) |
| Show More |
Safety and Handling
Signal Word
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?