missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteinase 3/Myeloblastin/PRTN3 Antibody (WGM2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11266
This item is not returnable.
View return policy
Description
Proteinase 3/Myeloblastin/PRTN3 Monoclonal specifically detects Proteinase 3/Myeloblastin/PRTN3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay.
Specifications
| Proteinase 3/Myeloblastin/PRTN3 | |
| Monoclonal | |
| 0.1 mg/ml | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Immunoassay | |
| ACPA, AGP7PR3, azurophil granule protein 7, C-ANCA, EC 3.4.21, EC 3.4.21.76, Leukocyte proteinase 3, MBN, MBT, myeloblastin, Neutrophil proteinase 4, NP4, NP-4, P29C-ANCA antigen, PR-3CANCA, proteinase 3, serine proteinase, neutrophil, Wegener autoantigen, Wegener granulomatosis autoantigen | |
| Monoclonal antibody WGM2 reacts with human proteinase 3 (PR3). | |
| 50 μg | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers | |
| 5657 | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Immunoassay | |
| WGM2 | |
| Unconjugated | |
| 0.2 um filtered solution in PBS, 0.1% BSA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction