missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Z Monoclonal Antibody (OTI4F1)
Protein Z Monoclonal Antibody (OTI4F1)
Brand: Invitrogen MA526806
This item is not returnable.
View return policy
Description
Specifications
| Protein Z | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22891 | |
| PROZ | |
| Human recombinant protein fragment corresponding to amino acids 115-400 of PROZ produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F1 | |
| Unconjugated | |
| PROZ | |
| protein Z, vitamin K dependent plasma glycoprotein; protein Z, vitamin K-dependent plasma glycoprotein; PROZ; PZ; vitamin K-dependent protein Z; vitamin K-dependent protein Z precursor variant 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8858 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction