missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Z Antibody (2B4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47452
This item is not returnable.
View return policy
Description
Protein Z Monoclonal specifically detects Protein Z in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Protein Z | |
| Monoclonal | |
| Unconjugated | |
| protein Z, vitamin K-dependent plasma glycoprotein, PZ, vitamin K-dependent protein Z | |
| Mouse | |
| 45 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Ascites |
| Western Blot, ELISA | |
| 2B4 | |
| Western Blot 1: 500-1:2000, ELISA 1:10000 | |
| PROZ | |
| Purified recombinant fragment of Protein Z expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 8858 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction