Learn More
Cytiva™ Protein Select™ Resin
Product Code. 30284848
Change view
Click to view available options
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
30284848 Each
30284842 Each
30284840 Each
30284828 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30284848 Supplier Cytiva™ Supplier No. 0045CNP01TA
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
An affinity chromatography resin for purifying recombinant proteins using the self-cleaving Cytiva™ Protein Select™ tag.
  • Infrared (IR) turbine rotor reflection ensures accurate and repeatable flow measurements, ideal for precision-critical applications
  • Robust and stable design with four screw gaps allows for secure and easy mounting in various system configurations
  • Constructed from USP Class VI PVDF material, offering excellent chemical resistance and compatibility with aggressive liquids
  • Supports gamma sterilization up to 50kGy, making it suitable for use in sterile environments and medical-grade processes
  • Delivers high-resolution square wave output for reliable signal processing and integration with control systems
  • Tubes can be sterilized up to 150°C (302°F), enabling safe use in high-temperature cleaning and sterilization protocols
  • Designed to reduce downtime and prevent cross-contamination, enhancing operational efficiency and product safety
  • Each sensor is pre-calibrated to achieve accuracy within <1%, minimizing setup time and ensuring consistent performance
TRUSTED_SUSTAINABILITY
Specifications
Type Flowmeter
Accuracy ±1%
Airflow Rate 0.03 to 2 L/min.
Case Material PVDF
Float Material PVDF
Gas Type All
Pressure Rating 362 psig
Includes 1/8 NPT Tubeholder System