missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Protein Precipitation Buffer MP

Code produit. 15830908
Change view
Click to view available options
Quantity:
100 mL (for 250 preps)
20 mL (for 50 preps)
Conditionnement:
100mL
20mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantity unitSize
15830908 100 mL (for 250 preps) 100mL
15819097 20 mL (for 50 preps) 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 15830908 Fournisseur Macherey-Nagel™ Code fournisseur 740407.100

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Additional protein precipitation buffer for use with NucleoSpin™ miRNA kits for the isolation of total RNA (small and large RNA) from diverse sample materials.

NucleoSpin™ miRNA Kit for small and large RNA isolation

  • Excellent total RNA recovery and purity by chaotropic salt lysis
  • Convenient extraction without phenol/chloroform
  • Support protocol for fractionation of RNA by size in two separate fractions
    • Small RNA (< 200 nt) and Large RNA (> 200 nt)
  • Additional isolation of total protein fraction ready to use for SDS-PAGE and Western blot analysis
  • NucleoSpin™ Filters for efficient sample homogenization
  • rDNase for efficient on-column removal of genomic DNA

Spécification

Chemical Name or Material Protein Precipitation Buffer MP
CAS 127-09-3
Color Colorless
For Use With (Application) RNA Isolation
For Use With (Equipment) NucleoSpin miRNA kit
pH 4.0 to 4.5
Quantity 100 mL (for 250 preps)
Odor Acetic
Solubility Information Soluble in Water
compliance-icons compliance-icons compliance-icons
Mention d'avertissement
  • Danger
Catégorie de danger
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Mention de danger
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Conseils de prudence
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.