missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Protein Precipitation Buffer MP

Product Code. 15819097
Click to view available options
Quantity:
100 mL (for 250 preps)
20 mL (for 50 preps)
Unit Size:
100mL
20mL
This item is not returnable. View return policy

Product Code. 15819097

Brand: Macherey-Nagel™ 740407.20

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Additional protein precipitation buffer for use with NucleoSpin™ miRNA kits for the isolation of total RNA (small and large RNA) from diverse sample materials.

NucleoSpin™ miRNA Kit for small and large RNA isolation

  • Excellent total RNA recovery and purity by chaotropic salt lysis
  • Convenient extraction without phenol/chloroform
  • Support protocol for fractionation of RNA by size in two separate fractions
    • Small RNA (< 200 nt) and Large RNA (> 200 nt)
  • Additional isolation of total protein fraction ready to use for SDS-PAGE and Western blot analysis
  • NucleoSpin™ Filters for efficient sample homogenization
  • rDNase for efficient on-column removal of genomic DNA

Specifications

Chemical Name or Material Protein Precipitation Buffer MP
CAS 127-09-3
Color Colorless
Concentration Sodium acetate, Zinc chloride
For Use With (Application) RNA Isolation
For Use With (Equipment) NucleoSpin miRNA kit
pH 4.0 to 4.5
Quantity 20 mL (for 50 preps)
Formulation Sodium acetate, Zinc chloride
Odor Acetic
Solubility Information Soluble in Water
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Buffer MP (20 mL)
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : zinc chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.