missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein phosphatase 1F Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35535-20ul
This item is not returnable.
View return policy
Description
Protein phosphatase 1F Polyclonal antibody specifically detects Protein phosphatase 1F in Human samples. It is validated for ELISA,Western Blot
Specifications
| Protein phosphatase 1F | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Ca(2+)/calmodulin-dependent protein kinase phosphatase, CaMKPaseCaM-kinase phosphatase, EC 3.1.3.16, FEM-2, hFEM-2, KIAA0015POPX2CAMKP, Partner of PIX 2, PP2C phosphatase, Protein fem-2 homolog, protein phosphatase 1F, protein phosphatase 1F (PP2C domain containing), protein phosphatase, Mg2+/Mn2+ dependent, 1F | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human Protein phosphatase 1F (NP_055449.1).,, Sequence:, MSSGAPQKSSPMASGAEETPGFLDTLLQDFPALLNPEDPLPWKAPGTVLSQEEVEGELAELAMGFLGSRKAPPPLAAALAHEAVSQLLQTDLSEFRKLPREEEEEEEDDDEEEKAPVTLLDAQSLAQSFFNRLWEVAGQWQKQVPLAARASQRQWLVSIHAIRNTRRKMEDRHVSLPSFNQLFGLSDPVN | |
| 20 μL | |
| Apoptosis, Protein Phosphatase | |
| 9647 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction