missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein phosphatase 1F Antibody (6D11), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42648MFV610
This item is not returnable.
View return policy
Description
Protein phosphatase 1F Monoclonal antibody specifically detects Protein phosphatase 1F in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Protein phosphatase 1F | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 6D11 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| Ca(2+)/calmodulin-dependent protein kinase phosphatase, CaMKPaseCaM-kinase phosphatase, EC 3.1.3.16, FEM-2, hFEM-2, KIAA0015POPX2CAMKP, Partner of PIX 2, PP2C phosphatase, Protein fem-2 homolog, protein phosphatase 1F, protein phosphatase 1F (PP2C domain containing), protein phosphatase, Mg2+/Mn2+ dependent, 1F | |
| Recombinant human Protein phosphatase 1F (1-454aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, Protein Phosphatase | |
| 9647 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction