missing translation for 'onlineSavingsMsg'
Learn More

Protein phosphatase 1F Antibody (2A9), Novus Biologicals™

Product Code. 18387198 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387198 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387198 Supplier Novus Biologicals Supplier No. H00009647M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Protein phosphatase 1F Monoclonal antibody specifically detects Protein phosphatase 1F in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein phosphatase 1F
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2A9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055449
Gene Alias Ca(2+)/calmodulin-dependent protein kinase phosphatase, CaMKPaseCaM-kinase phosphatase, EC 3.1.3.16, FEM-2, hFEM-2, KIAA0015POPX2CAMKP, Partner of PIX 2, PP2C phosphatase, Protein fem-2 homolog, protein phosphatase 1F, protein phosphatase 1F (PP2C domain containing), protein phosphatase, Mg2+/Mn2+ dependent, 1F
Host Species Mouse
Immunogen PPM1F (NP_055449, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSSGAPQKSSPMASGAEETPGFLDTLLQDFPALLNPEDPLPWKAPGTVLSQEEVEGELAELAMGFLGSRKAPPPLAAALAHEAVSQLLQTDLSEFRKLPR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 9647
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.