missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Phosphatase 1C gamma Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Protein Phosphatase 1C gamma Polyclonal specifically detects Protein Phosphatase 1C gamma in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
Specifications
| Antigen | Protein Phosphatase 1C gamma |
| Applications | ChIP Assay, Immunohistochemistry, KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | EC 3.1.3.16, PP-1G, PP1gamma, PPP1G, protein phosphatase 1, catalytic subunit, gamma isoform, protein phosphatase 1, catalytic subunit, gamma isozyme, Protein phosphatase 1C catalytic subunit, serine/threonine phosphatase 1 gamma, serine/threonine-protein phosphatase PP1-gamma catalytic subunit |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-323 of human Protein Phosphatase 1C gamma (NP_002701.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?