missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Protein Phosphatase 1C gamma Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Protein Phosphatase 1C gamma |
|---|---|
| Dilution | Western Blot : 10 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066807
|
Novus Biologicals™
NBP3-30770-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067095
|
Novus Biologicals™
NBP3-30770-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Protein Phosphatase 1C gamma Polyclonal antibody specifically detects Protein Phosphatase 1C gamma in Bovine,Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| Protein Phosphatase 1C gamma | |
| Western Blot | |
| Unconjugated | |
| Sheep | |
| Cancer, Lipid and Metabolism, Protein Phosphatase, Stem Cell Markers | |
| Phosphate buffered saline | |
| 5501 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 10 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine, Human, Mouse, Rat | |
| EC 3.1.3.16, PP-1G, PP1gamma, PPP1G, protein phosphatase 1, catalytic subunit, gamma isoform, protein phosphatase 1, catalytic subunit, gamma isozyme, Protein phosphatase 1C catalytic subunit, serine/threonine phosphatase 1 gamma, serine/threonine-protein phosphatase PP1-gamma catalytic subunit | |
| QKASNYRNNTVKYE | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title