missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Phosphatase 1 beta Antibody (8A7C7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52539
This item is not returnable.
View return policy
Description
Protein Phosphatase 1 beta Monoclonal specifically detects Protein Phosphatase 1 beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| Protein Phosphatase 1 beta | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| EC 3.1.3.16, EC 3.1.3.53, MGC3672, PP1beta, PP-1Bprotein phosphatase 1, catalytic subunit, delta isoform, PPP1CD, protein phosphatase 1, catalytic subunit, beta isoform, protein phosphatase 1, catalytic subunit, beta isozyme, protein phosphatase 1-beta, protein phosphatase 1-delta, serine/threonine protein phosphatase PP1-beta catalytic subunit, serine/threonine-protein phosphatase PP1-beta catalytic subunit | |
| Mouse | |
| 37.2 kDa | |
| 0.1 mg | |
| Cancer, Lipid and Metabolism, Protein Phosphatase | |
| 5500 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 8A7C7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PPP1CB | |
| Purified recombinant fragment of human PPP1CB (AA: 174-327) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction