missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 Antibody [DyLight 650], Novus Biologicals™
Shop All Bio Techne ProductsDescription
Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 Polyclonal antibody specifically detects Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Protein O-Glucosyltransferase 1/POGLUT1/KTELC1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 9630046K23Rik, C3orf9, CAP10-like protein, 46 kDa, chromosome 3 open reading frame 9, CLP46, EC 2.4.1.-, hCLP46CAP10-like 46 kDa protein, KDELC family like 1, KDELCL1, KTEL (Lys-Tyr-Glu-Leu) containing 1, KTEL motif-containing protein 1, MDS010, MDSRPKTELC1, MGC32995, Myelodysplastic syndromes relative protein, protein O-glucosyltransferase 1, Rumi, x 010 protein |
| Host Species | Sheep |
| Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Protein O-Glucosyltransferase 1/POGLUT1, Arg24-Leu388, Accession # Q8NBL1 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?