missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein mab-21-like 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19961
This item is not returnable.
View return policy
Description
Protein mab-21-like 1 Polyclonal specifically detects Protein mab-21-like 1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Protein mab-21-like 1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| CAGR1mab-21 (C. elegans)-like 1, FLJ10197, mab-21-like 1 (C. elegans), mab-21-like protein 1, protein mab-21-like 1 | |
| Rabbit | |
| 41 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rat (100%), Bovine (100%), Chicken (100%), Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.89mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q13394 | |
| MAB21L1 | |
| Recombinant protein encompassing a sequence within the center region of human MAB21L1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 4081 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction