missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Protein L Polyclonal Antibody

Chicken Polyclonal Antibody
Brand: Invitrogen™ PA172066
This item is not returnable.
View return policy
Description
PA1-72066 detects Protein L in bacteria samples. PA1-72066 has been successfully used in ELISA and western blot procedures. The PA1-72066 immunogen is from purified recombinant protein L.
Specifications
| Protein L | |
| Polyclonal | |
| Unconjugated | |
| Chicken | |
| Affinity Chromatography | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgY |
| ELISA, Western Blot | |
| 1.0 mg/mL | |
| PBS with 0.075% sodium azide; pH 7.2 | |
| Purified recombinant protein L. | |
| 500 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction