missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein kinase Y linked Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00005616-D01P
This item is not returnable.
View return policy
Description
Protein kinase Y linked Polyclonal antibody specifically detects Protein kinase Y linked in Human samples. It is validated for Western Blot
Specifications
| Protein kinase Y linked | |
| Polyclonal | |
| Western Blot | |
| EC 2.7.11, EC 2.7.11.1, protein kinase, Y-linked | |
| PRKY (NP_002751.1, 1 a.a. - 277 a.a.) full-length human protein. MEAPGPAQAAAAESNSREVTEDAADWAPALCPSPEARSPEAPAYRLQDCDALVTMGTGTFGRVHLVKEKTAKHFFALKVMSIPDVIRRKQEQHVHNEKSVLKEVSHPFLIRLFWTWHEERFLYMLMEYVPGGELFSYLRNRGHFSSTTGLFYSAEIICAIEYLHSKEIVYRDLKPENILLDRDGHIKLTDFGFAKKLVDRTWTLCGTPEYLAPEVIQSKGHGRAVDWWALGILIFEMLSGFPPFFDDNPFGIYQKILAGKLYFPRHLDFHVKTGRMM | |
| 0.1 mg | |
| Protein Kinase, Wnt Signaling Pathway | |
| 5616 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction