missing translation for 'onlineSavingsMsg'
Learn More

Protein kinase Y linked Antibody, Novus Biologicals™

Product Code. 18366938 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366938 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366938 Supplier Novus Biologicals Supplier No. H00005616B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Protein kinase Y linked Polyclonal antibody specifically detects Protein kinase Y linked in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein kinase Y linked
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Alias EC 2.7.11, EC 2.7.11.1, protein kinase, Y-linked
Host Species Mouse
Immunogen PRKY (NP_002751.1, 1 a.a. - 277 a.a.) full-length human protein. MEAPGPAQAAAAESNSREVTEDAADWAPALCPSPEARSPEAPAYRLQDCDALVTMGTGTFGRVHLVKEKTAKHFFALKVMSIPDVIRRKQEQHVHNEKSVLKEVSHPFLIRLFWTWHEERFLYMLMEYVPGGELFSYLRNRGHFSSTTGLFYSAEIICAIEYLHSKEIVYRDLKPENILLDRDGHIKLTDFGFAKKLVDRTWTLCGTPEYLAPEVIQSKGHGRAVDWWALGILIFEMLSGFPPFFDDNPFGIYQKILAGKLYFPRHLDFHVKTGRMM
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5616
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.