missing translation for 'onlineSavingsMsg'
Learn More

Protein Kinase D2 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18368726 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368726 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368726 Supplier Novus Biologicals Supplier No. NBP299858DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Protein Kinase D2 Polyclonal specifically detects Protein Kinase D2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein Kinase D2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias DKFZP586E0820, EC 2.7.11, EC 2.7.11.13, nPKC-D2, PKD2HSPC187, protein kinase D2, serine/threonine-protein kinase D2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Protein Kinase D2 (NP_057541.2; Met1-Leu878).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Phospho Specific, Protein Kinase, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 25865
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.