missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control

Product Code. 15440597
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440597 1 mg 1mg
15430597 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440597 Supplier Thermo Scientific Chemicals Supplier No. J66521.M^
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For cellular and molecular biology applications

A scrambled peptide with an identical amino acid composition to that of Protein Kinase C epsilon translocation Inhibitor Peptide. A cell impermeable negative control for PKC translocation inhibition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
For Use With (Application) A scrambled peptide with an identical amino acid composition to that of Protein Kinase C epsilon translocation Inhibitor Peptide. A cell impermeable negative control for PKC translocation inhibition.
Molecular Weight (g/mol) 843.98
Product Type Protein Kinase C-epsilon Translocation Inhibitor Peptide, Negative Control
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence Leu-Ser-Glu-Thr-Lys-Pro-Ala-Val

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.