missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Protein Kinase A Regulatory Subunit II alpha Rabbit anti-Human, Clone: 9C6B3, Novus Biologicals™
Description
Protein Kinase A Regulatory Subunit II alpha Monoclonal antibody specifically detects Protein Kinase A Regulatory Subunit II alpha in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Protein Kinase A Regulatory Subunit II alpha |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 9C6B3 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 312-404 of human Protein Kinase A Regulatory Subunit II alpha (P13861). |
| Purification Method | Affinity purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?