missing translation for 'onlineSavingsMsg'
Learn More

Protein Kinase A regulatory subunit I alpha Rabbit anti-Human, Mouse, Rat, Clone: S06-8A5, Novus Biologicals™

Product Code. 18373532
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373532 25 μg 25µL
18394692 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18373532 Supplier Bio-Techne Supplier No. NBP31508625UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Protein Kinase A regulatory subunit I alpha Monoclonal antibody specifically detects Protein Kinase A regulatory subunit I alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein Kinase A regulatory subunit I alpha
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S06-8A5
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias cAMP-dependent protein kinase regulatory subunit RIalpha, cAMP-dependent protein kinase type I-alpha regulatory chain, cAMP-dependent protein kinase type I-alpha regulatory subunit, CAR, CNC, CNC1, MGC17251, PKA R1 alpha, PKA1, PKR1, PPNAD1, PRKAR1, protein kinase A type 1a regulatory subunit, protein kinase, cAMP-dependent, regulatory, type I, alpha (tissue specificextinguisher 1), Tissue-specific extinguisher 1, TSE1DKFZp779L0468
Host Species Rabbit
Immunogen A synthetic peptide of human Protein Kinase A regulatory subunit I alpha (Uniprot#: P10644)
Purification Method Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5573
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.