missing translation for 'onlineSavingsMsg'
Learn More

Protein Kinase A regulatory subunit I alpha Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-93193-0.1ml

 View more versions of this product

Product Code. 18627521

  • £375.00 / 0.01mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Protein Kinase A regulatory subunit I alpha Polyclonal antibody specifically detects Protein Kinase A regulatory subunit I alpha in Human samples. It is validated for Western Blot, Gene Knock-Out
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Protein Kinase A regulatory subunit I alpha
Polyclonal
Western Blot 1:500 - 1:1000, Knockout Validated
cAMP-dependent protein kinase regulatory subunit RIalpha, cAMP-dependent protein kinase type I-alpha regulatory chain, cAMP-dependent protein kinase type I-alpha regulatory subunit, CAR, CNC, CNC1, MGC17251, PKA R1 alpha, PKA1, PKR1, PPNAD1, PRKAR1, protein kinase A type 1a regulatory subunit, protein kinase, cAMP-dependent, regulatory, type I, alpha (tissue specificextinguisher 1), Tissue-specific extinguisher 1, TSE1DKFZp779L0468
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human Protein Kinase A regulatory subunit I alpha (NP_002725.1). MESGSTAASEEARSLRECELYVQKHNIQALLKDSIVQLCTARPERPMAFLREYFERLEKEEAKQIQNLQKAGTRTDSREDEISPPPPNPVVKGRRRRGAISAEVYTEEDAASYVRKVIPKDYKTMAALAKAIEKNVLFSHLDDNERSDIFDAMFSVSFIAGETVIQQGDEGDNFYVIDQGETDVYVNNEWATSVGEGGSFGELALIYGTPRAATVKAKTNVKLWGIDRDSYRRILMGSTLRKRKMYEEFL
0.1 mL
Cancer
5573
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Gene Knock-Out
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.