missing translation for 'onlineSavingsMsg'
Learn More

Protein Kinase A regulatory subunit I alpha Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18329222
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329222 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329222 Supplier Bio-Techne Supplier No. NBP306561AF700

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Protein Kinase A regulatory subunit I alpha Polyclonal antibody specifically detects Protein Kinase A regulatory subunit I alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein Kinase A regulatory subunit I alpha
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias cAMP-dependent protein kinase regulatory subunit RIalpha, cAMP-dependent protein kinase type I-alpha regulatory chain, cAMP-dependent protein kinase type I-alpha regulatory subunit, CAR, CNC, CNC1, MGC17251, PKA R1 alpha, PKA1, PKR1, PPNAD1, PRKAR1, protein kinase A type 1a regulatory subunit, protein kinase, cAMP-dependent, regulatory, type I, alpha (tissue specificextinguisher 1), Tissue-specific extinguisher 1, TSE1DKFZp779L0468
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Protein Kinase A regulatory subunit I alpha fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5573
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.