missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Protein Gene Product 9.5 (PGP9.5) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Epredia™ Protein Gene Product 9.5 (PGP9.5) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1760-S1

 View more versions of this product

Product Code. 12600127

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Protein gene product 9.5 is a ubiquitin C-terminal hydroxylase found in neurons and nerve fibres of central and peripheral nervous system; many neuroendocrinal cells, segments of renal tubules, spermatogonia, leydig cells of testis; and ova. It is present in cellular inclusions and can be a useful as a neuronal marker and in the studies of neurodegenerative disorders.

Host Species: Mouse

Clone: 10A1

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant protein corresponding to full-length human protein gene product 9.5 molecule

Molecular Weight: 25-27kDa

Positive Control: Small intestine

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Protein Gene Product 9.5 (PGP9.5) Ab-1
Monoclonal
Unconjugated
Recombinant protein corresponding to full length human protein gene product 95 molecule
500 μL
Neuroscience
Human
IgG2b
Immunohistochemistry (Paraffin)
10A1
Mouse
Small intestine
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.